Assay Details
| Gene Symbol: LOC4326755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765686.1 | XP_015621172.1 | 2 - 3 | 395 | 134 | |
| GenBank mRNA | AK120732.1 | - | 2 - 3 | 306 | 134 | |
More Information
Set Membership: |
|

LOC4326755
Genomic Map