Assay Details
Gene Symbol: LOC4325165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015796015.2 | XP_015651501.1 | 1 - 2 | 215 | 109 | |
GenBank mRNA | AK071543.1 | - | 1 - 2 | 185 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015796015.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
