Assay Details
Gene Symbol: LOC4325165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755116.2 | XP_015610602.1 | 4 - 5 | 752 | 59 | |
XM_015755124.2 | XP_015610610.1 | 4 - 5 | 755 | 59 | ||
XM_015795996.2 | XP_015651482.1 | 4 - 5 | 758 | 59 | ||
XM_015796006.2 | XP_015651492.1 | 5 - 6 | 725 | 59 | ||
XM_015796015.2 | XP_015651501.1 | 5 - 6 | 721 | 59 | ||
GenBank mRNA | AK071543.1 | - | 5 - 6 | 691 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
