Assay Details
| Gene Symbol: LOC4327755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768197.2 | XP_015623683.1 | 4 - 5 | 826 | 117 | |
| GenBank mRNA | AK059447.1 | - | 5 - 6 | 779 | 117 | |
| AK099426.1 | - | 4 - 5 | 802 | 117 | ||
More Information
Set Membership: |
|

LOC4327755
Genomic Map