Assay Details
| Gene Symbol: LOC4326283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772035.2 | XP_015627521.1 | 8 - 9 | 1543 | 106 | |
| GenBank mRNA | AK120976.1 | - | 8 - 9 | 1472 | 106 | |
| Y11189.1 | - | 8 - 9 | 702 | 106 | ||
More Information
Set Membership: |
|

LOC4326283
Genomic Map