Assay Details
| Gene Symbol: LOC4326596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794946.2 | XP_015650432.1 | 5 - 6 | 862 | 71 | |
| GenBank mRNA | AK108988.1 | - | 5 - 6 | 700 | 71 | |
More Information
Set Membership: |
|

LOC4326596
Genomic Map