Assay Details
Gene Symbol: LOC4326555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766979.2 | XP_015622465.1 | 1 - 2 | 420 | 74 | |
GenBank mRNA | AK243497.1 | - | 1 - 2 | 379 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
