Assay Details
Gene Symbol: LOC4326555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766979.2 | XP_015622465.1 | 4 - 5 | 691 | 84 | |
GenBank mRNA | AK243497.1 | - | 4 - 5 | 650 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
