Assay Details
Gene Symbol: LOC4326555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766979.2 | XP_015622465.1 | 7 - 8 | 857 | 81 | |
GenBank mRNA | AK243497.1 | - | 7 - 8 | 816 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
