Assay Details
| Gene Symbol: LOC4325922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768570.2 | XP_015624056.1 | 1 | 1330 | 112 | |
| GenBank mRNA | AK242412.1 | - | 1 | 1179 | 112 | |
More Information
Set Membership: |
|

LOC4325922
Genomic Map