Assay Details
Gene Symbol: LOC4325365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790350.2 | XP_015645836.1 | 3 - 4 | 705 | 80 | |
GenBank mRNA | AK242032.1 | - | 3 - 4 | 712 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
