Assay Details
| Gene Symbol: LOC4325365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790350.2 | XP_015645836.1 | 4 - 5 | 983 | 58 | |
| GenBank mRNA | AK242032.1 | - | 4 - 5 | 990 | 58 | |
More Information
Set Membership: |
|

LOC4325365
Genomic Map