Assay Details
Gene Symbol: LOC4325619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015773108.2 | 1 - 2 | 221 | 63 | ||
GenBank mRNA | AK070142.1 | - | 1 - 2 | 225 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773108.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
