Assay Details
| Gene Symbol: LOC4327535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765657.2 | XP_015621143.1 | 1 - 2 | 355 | 61 | |
| GenBank mRNA | AF435976.1 | - | 1 - 2 | 198 | 61 | |
| AK061770.1 | - | 1 - 2 | 357 | 61 | ||
| AK072206.1 | - | 1 - 2 | 220 | 61 | ||
| AK121632.1 | - | 1 - 2 | 220 | 61 | ||
| BT129032.1 | - | 1 - 2 | 351 | 61 | ||
| BT129753.1 | - | 1 - 2 | 214 | 61 | ||
| BT131286.1 | - | 1 - 2 | 218 | 61 | ||
| KM982768.1 | - | 1 - 2 | 321 | 61 | ||
| KM982769.1 | - | 1 - 2 | 181 | 61 | ||
| Gene Symbol: LOC9271026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK241277.1 | - | 220 | 61 | ||
More Information
Set Membership: |
|

LOC4327535
Genomic Map