Assay Details
Gene Symbol: LOC4324888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765696.2 | XP_015621182.1 | 1 - 2 | 175 | 120 | |
GenBank mRNA | AK070958.1 | - | 1 - 2 | 176 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765696.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
