Assay Details
Gene Symbol: LOC4324888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765696.2 | XP_015621182.1 | 3 - 4 | 454 | 81 | |
GenBank mRNA | AK070958.1 | - | 3 - 4 | 455 | 81 | |
AY551913.1 | - | 2 - 3 | 345 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
