Assay Details
| Gene Symbol: LOC4324888 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765696.2 | XP_015621182.1 | 4 - 5 | 529 | 67 | |
| GenBank mRNA | AK070958.1 | - | 4 - 5 | 530 | 67 | |
| AY551913.1 | - | 3 - 4 | 420 | 67 | ||
More Information
Set Membership: |
|

LOC4324888
Genomic Map