Assay Details
Gene Symbol: LOC4324979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766903.2 | XP_015622389.1 | 10 - 11 | 1649 | 93 | |
GenBank mRNA | AK103626.1 | - | 10 - 11 | 1614 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
