Assay Details
Gene Symbol: LOC4324979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766903.2 | XP_015622389.1 | 11 - 12 | 1722 | 59 | |
GenBank mRNA | AK103626.1 | - | 11 - 12 | 1687 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
