Assay Details
Gene Symbol: LOC4324330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761170.2 | XP_015616656.1 | 1 - 2 | 1171 | 78 | |
GenBank mRNA | AK109173.1 | - | 1 - 2 | 42 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761170.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
