Assay Details
| Gene Symbol: LOC4326815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765771.2 | XP_015621257.1 | 1 - 2 | 446 | 91 | |
| XM_015765772.2 | XP_015621258.1 | 1 - 2 | 344 | 91 | ||
| GenBank mRNA | AK059213.1 | - | 1 - 2 | 206 | 91 | |
| AK068780.1 | - | 1 - 2 | 359 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765772.2 transcripts.
Set Membership: |
|

LOC4326815
Genomic Map