Assay Details
Gene Symbol: LOC4326815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765771.2 | XP_015621257.1 | 5 - 6 | 974 | 87 | |
XM_015765772.2 | XP_015621258.1 | 5 - 6 | 883 | 87 | ||
GenBank mRNA | AK059213.1 | - | 5 - 6 | 734 | 87 | |
AK068780.1 | - | 5 - 6 | 887 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
