Assay Details
Gene Symbol: LOC4326815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765771.2 | XP_015621257.1 | 7 - 8 | 1184 | 71 | |
XM_015765772.2 | XP_015621258.1 | 7 - 8 | 1093 | 71 | ||
GenBank mRNA | AK059213.1 | - | 7 - 8 | 944 | 71 | |
AK068780.1 | - | 7 - 8 | 1097 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
