Assay Details
Gene Symbol: LOC4326815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765771.2 | XP_015621257.1 | 8 - 9 | 1259 | 68 | |
XM_015765772.2 | XP_015621258.1 | 8 - 9 | 1168 | 68 | ||
GenBank mRNA | AK059213.1 | - | 8 - 9 | 1019 | 68 | |
AK068780.1 | - | 8 - 9 | 1172 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
