Assay Details
| Gene Symbol: LOC4326524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761305.2 | XP_015616791.1 | 7 - 8 | 690 | 89 | |
| GenBank mRNA | AK101890.1 | - | 7 - 8 | 682 | 89 | |
More Information
Set Membership: |
|

LOC4326524
Genomic Map