Assay Details
| Gene Symbol: LOC4328183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767273.2 | XP_015622759.1 | 2 - 3 | 736 | 109 | |
| GenBank mRNA | AK120049.1 | - | 2 - 3 | 725 | 109 | |
More Information
Set Membership: |
|

LOC4328183
Genomic Map