Assay Details
Gene Symbol: LOC4329037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771915.2 | 5 - 6 | 889 | 84 | ||
GenBank mRNA | AK060415.1 | - | 2 - 3 | 206 | 84 | |
AK099035.1 | - | 5 - 6 | 889 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
