Assay Details
Gene Symbol: LOC4329037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771915.2 | 6 - 7 | 972 | 139 | ||
GenBank mRNA | AK060415.1 | - | 3 - 4 | 289 | 139 | |
AK072177.1 | - | 6 - 7 | 977 | 139 | ||
AK099035.1 | - | 6 - 7 | 972 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
