Assay Details
| Gene Symbol: LOC112936002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771666.2 | XP_015627152.1 | 2 - 3 | 423 | 112 | |
| XM_015771667.2 | XP_015627153.1 | 2 - 3 | 1038 | 112 | ||
| GenBank mRNA | AK070515.1 | - | 2 - 3 | 333 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771666.2, XM_015771667.2 transcripts.
Set Membership: |
|

LOC112936002
Genomic Map