Assay Details
| Gene Symbol: LOC112936002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771663.2 | XP_015627149.1 | 6 - 7 | 866 | 95 | |
| XM_015771664.2 | XP_015627150.1 | 7 - 8 | 771 | 95 | ||
| XM_015771665.2 | XP_015627151.1 | 6 - 7 | 1483 | 95 | ||
| XM_015771666.2 | XP_015627152.1 | 6 - 7 | 856 | 95 | ||
| XM_015771667.2 | XP_015627153.1 | 6 - 7 | 1471 | 95 | ||
| GenBank mRNA | AK070515.1 | - | 6 - 7 | 766 | 95 | |
More Information
Set Membership: |
|

LOC112936002
Genomic Map