Assay Details
| Gene Symbol: LOC4329546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767708.2 | XP_015623194.1 | 13 - 14 | 2192 | 66 | |
| GenBank mRNA | AK102681.1 | - | 13 - 14 | 1962 | 66 | |
| AK105277.1 | - | 1 - 2 | 154 | 66 | ||
More Information
Set Membership: |
|

LOC4329546
Genomic Map