Assay Details
| Gene Symbol: LOC4329678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771102.2 | XP_015626588.1 | 3 - 4 | 824 | 76 | |
| XM_015771103.2 | XP_015626589.1 | 4 - 5 | 867 | 76 | ||
| XM_015771104.2 | XP_015626590.1 | 4 - 5 | 857 | 76 | ||
| XM_015771105.2 | XP_015626591.1 | 3 - 4 | 819 | 76 | ||
| XM_015771106.2 | XP_015626592.1 | 3 - 4 | 819 | 76 | ||
| XM_015771107.2 | XP_015626593.1 | 3 - 4 | 702 | 76 | ||
| GenBank mRNA | AK069669.1 | - | 3 - 4 | 695 | 76 | |
More Information
Set Membership: |
|

LOC4329678
Genomic Map