Assay Details
| Gene Symbol: LOC4329678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771102.2 | XP_015626588.1 | 5 - 6 | 1271 | 86 | |
| XM_015771103.2 | XP_015626589.1 | 6 - 7 | 1314 | 86 | ||
| XM_015771104.2 | XP_015626590.1 | 6 - 7 | 1304 | 86 | ||
| XM_015771105.2 | XP_015626591.1 | 5 - 6 | 1266 | 86 | ||
| XM_015771106.2 | XP_015626592.1 | 5 - 6 | 1266 | 86 | ||
| XM_015771107.2 | XP_015626593.1 | 5 - 6 | 1149 | 86 | ||
| GenBank mRNA | AB069967.1 | - | 4 - 5 | 753 | 86 | |
| AK066353.1 | - | 5 - 6 | 1291 | 86 | ||
| AK069669.1 | - | 5 - 6 | 1142 | 86 | ||
More Information
Set Membership: |
|

LOC4329678
Genomic Map