Assay Details
| Gene Symbol: LOC4329744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767277.2 | XP_015622763.1 | 3 - 4 | 778 | 87 | |
| XM_015767278.2 | XP_015622764.1 | 3 - 4 | 786 | 87 | ||
| GenBank mRNA | AK065365.1 | - | 3 - 4 | 620 | 87 | |
| AK103980.1 | - | 3 - 4 | 517 | 87 | ||
More Information
Set Membership: |
|

LOC4329744
Genomic Map