Assay Details
| Gene Symbol: LOC4329744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767277.2 | XP_015622763.1 | 4 - 5 | 1420 | 66 | |
| XM_015767278.2 | XP_015622764.1 | 4 - 5 | 1428 | 66 | ||
| GenBank mRNA | AK059863.1 | - | 2 - 3 | 687 | 66 | |
| AK065365.1 | - | 4 - 5 | 1262 | 66 | ||
| AK103980.1 | - | 4 - 5 | 1159 | 66 | ||
More Information
Set Membership: |
|

LOC4329744
Genomic Map