Assay Details
Gene Symbol: LOC4329857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767643.2 | XP_015623129.1 | 1 | 681 | 75 | |
GenBank mRNA | AK069935.1 | - | 1 | 655 | 75 | |
AK099175.1 | - | 1 | 654 | 75 | ||
AK103993.1 | - | 1 | 653 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
