Assay Details
| Gene Symbol: LOC4329962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769820.2 | XP_015625306.1 | 1 | 1796 | 124 | |
| GenBank mRNA | AK110995.1 | - | 1 | 1659 | 124 | |
More Information
Set Membership: |
|

LOC4329962
Genomic Map