Assay Details
| Gene Symbol: LOC4329963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769822.2 | XP_015625308.1 | 2 - 3 | 478 | 57 | |
| GenBank mRNA | AK061262.1 | - | 2 - 3 | 431 | 57 | |
| AK069360.1 | - | 2 - 3 | 421 | 57 | ||
| AK101546.1 | - | 2 - 3 | 432 | 57 | ||
| AK288426.1 | - | 2209 | 57 | |||
| AY278997.1 | - | 2 - 3 | 249 | 57 | ||
More Information
Set Membership: |
|

LOC4329963
Genomic Map