Assay Details
| Gene Symbol: LOC4329963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769822.2 | XP_015625308.1 | 3 - 4 | 608 | 74 | |
| GenBank mRNA | AK061262.1 | - | 3 - 4 | 561 | 74 | |
| AK069360.1 | - | 3 - 4 | 551 | 74 | ||
| AK101546.1 | - | 3 - 4 | 562 | 74 | ||
| AK288426.1 | - | 2339 | 74 | |||
| AY278997.1 | - | 3 - 4 | 379 | 74 | ||
More Information
Set Membership: |
|

LOC4329963
Genomic Map