Assay Details
| Gene Symbol: LOC4329963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769822.2 | XP_015625308.1 | 5 - 6 | 741 | 98 | |
| GenBank mRNA | AK069360.1 | - | 5 - 6 | 684 | 98 | |
| AK101546.1 | - | 5 - 6 | 695 | 98 | ||
| AK288426.1 | - | 2472 | 98 | |||
| AY278997.1 | - | 5 - 6 | 512 | 98 | ||
More Information
Set Membership: |
|

LOC4329963
Genomic Map