Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 11 - 12 | 1291 | 97 | |
| XM_015770625.1 | XP_015626111.1 | 11 - 12 | 1290 | 97 | ||
| XM_015770627.1 | XP_015626113.1 | 10 - 11 | 779 | 97 | ||
| XM_015770628.2 | XP_015626114.1 | 11 - 12 | 1254 | 97 | ||
| GenBank mRNA | AK065045.1 | - | 11 - 12 | 1256 | 97 | |
More Information
Set Membership: |
|

LOC4330499
Genomic Map