Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 17 - 18 | 2381 | 66 | |
| XM_015770625.1 | XP_015626111.1 | 17 - 18 | 2380 | 66 | ||
| XM_015770627.1 | XP_015626113.1 | 16 - 17 | 1869 | 66 | ||
| XM_015770628.2 | XP_015626114.1 | 17 - 18 | 2344 | 66 | ||
| GenBank mRNA | AK065045.1 | - | 17 - 18 | 2346 | 66 | |
More Information
Set Membership: |
|

LOC4330499
Genomic Map