Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 6 - 7 | 907 | 78 | |
| XM_015770625.1 | XP_015626111.1 | 6 - 7 | 906 | 78 | ||
| XM_015770627.1 | XP_015626113.1 | 5 - 6 | 395 | 78 | ||
| XM_015770628.2 | XP_015626114.1 | 6 - 7 | 870 | 78 | ||
| GenBank mRNA | AK065045.1 | - | 6 - 7 | 872 | 78 | |
More Information
Set Membership: |
|

LOC4330499
Genomic Map