Assay Details
| Gene Symbol: LOC4330560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767435.1 | XP_015622921.1 | 2 - 3 | 511 | 126 | |
| GenBank mRNA | AK061514.1 | - | 2 - 3 | 423 | 126 | |
| AK104061.1 | - | 2 - 3 | 361 | 126 | ||
More Information
Set Membership: |
|

LOC4330560
Genomic Map