Assay Details
| Gene Symbol: LOC4330747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767928.2 | XP_015623414.1 | 6 - 7 | 1134 | 95 | |
| GenBank mRNA | AK101802.1 | - | 6 - 7 | 1018 | 95 | |
More Information
Set Membership: |
|

LOC4330747
Genomic Map