Assay Details
Gene Symbol: LOC4331761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775251.2 | XP_015630737.1 | 1 - 2 | 259 | 51 | |
GenBank mRNA | AK061772.1 | - | 1 - 2 | 162 | 51 | |
AK066306.1 | - | 1 - 2 | 179 | 51 | ||
AK099215.1 | - | 1 - 2 | 146 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
