Assay Details
Gene Symbol: LOC4331761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775251.2 | XP_015630737.1 | 4 - 5 | 622 | 70 | |
GenBank mRNA | AK061772.1 | - | 4 - 5 | 525 | 70 | |
AK066306.1 | - | 4 - 5 | 542 | 70 | ||
AK099215.1 | - | 4 - 5 | 509 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
