Assay Details
| Gene Symbol: LOC4331761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775251.2 | XP_015630737.1 | 6 - 7 | 809 | 80 | |
| GenBank mRNA | AK061772.1 | - | 6 - 7 | 712 | 80 | |
| AK066306.1 | - | 6 - 7 | 729 | 80 | ||
| AK099215.1 | - | 6 - 7 | 696 | 80 | ||
More Information
Set Membership: |
|

LOC4331761
Genomic Map