Assay Details
| Gene Symbol: LOC4331761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775251.2 | XP_015630737.1 | 7 - 8 | 865 | 84 | |
| GenBank mRNA | AK061772.1 | - | 7 - 8 | 768 | 84 | |
| AK066306.1 | - | 7 - 8 | 785 | 84 | ||
| AK099215.1 | - | 7 - 8 | 752 | 84 | ||
More Information
Set Membership: |
|

LOC4331761
Genomic Map