Assay Details
Gene Symbol: LOC4332700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772956.2 | XP_015628442.1 | 6 - 7 | 618 | 94 | |
GenBank mRNA | AK059868.1 | - | 6 - 7 | 604 | 94 | |
AK071431.1 | - | 6 - 7 | 600 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
